Learn More
3-Chlorobenzyl Cyanide, TRC
Product Code. 30367919
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367919 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30367919 Supplier Toronto Research Chemicals Supplier No. TRCC36445550G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Chlorobenzyl Cyanide
CAS 1529-41-5
Molecular Formula C8 H6 Cl N
Quantity 50 g
Synonym 3-Chloro-benzeneacetonitrile, (m-Chlorophenyl)-acetonitrile, 1-Chloro-3-(cyanomethyl)benzene, 2-(3-Chlorophenyl)acetonitrile, 3-Chlorobenzeneacetonitrile, 3-Chlorobenzyl cyanide, 3-Chlorophenylacetonitrile, m-Chlorobenzyl Cyanide, m-Chlorobenzyl Nitrile, m-Chlorophenylacetonitrile
InChI Formula InChI=1S/C8H6ClN/c9-8-3-1-2-7(6-8)4-5-10/h1-3,6H,4H2
SMILES Clc1cccc(CC#N)c1
IUPAC Name 2-(3-chlorophenyl)acetonitrile
Molecular Weight (g/mol) 151.59
Formula Weight 151.0189