missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorocarbonylbenzeneboronic anhydride, tech. 90%

Product Code. 15430066
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430066 1 g 1g
15440066 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15430066 Supplier Thermo Scientific Alfa Aesar Supplier No. H52722.03
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 332154-58-2 | C7H6BClO3 | 184.382 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332154-58-2
Molecular Formula C7H6BClO3
Molecular Weight (g/mol) 184.382
MDL Number MFCD06801675
InChI Key AIHGXUQQIIYAJP-UHFFFAOYSA-N
Synonym 3-chlorocarbonylphenylboronic acid, 3-chlorocarbonyl phenyl boronic acid, benzoylchloride-3-boronic acid, 3-carbonochloridoyl phenylboronic acid, pubchem9524, acmc-209hzb, 3-chlorocarbonylphenylboronicacid, boronic acid, 3-chlorocarbonyl phenyl-9ci
PubChem CID 17750521
IUPAC Name (3-carbonochloridoylphenyl)boronic acid
SMILES B(C1=CC(=CC=C1)C(=O)Cl)(O)O

Specifications

Melting Point 252°C to 254°C
Quantity 1 g
Formula Weight 166.37
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-Chlorocarbonylbenzeneboronic anhydride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.