missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloroperbenzoic Acid (Technical Grade, wet with water), TRC
Click to view available options
Quantity:
100 g
Unit Size:
100g
Specifications
Specifications
| Chemical Name or Material | 3-Chloroperbenzoic Acid (Technical Grade) |
| CAS | 937-14-4 |
| Molecular Formula | C7H5ClO3 |
| Quantity | 100 g |
| Synonym | 3-Chlorobenzenecarboperoxoic Acid, m-Chloroperoxybenzoic Acid, 3-Chlorobenzenecarboperoxoic Acid, 3-Chloroperbenzoic Acid, 3-Chloroperoxybenzoic Acid, MCPBA, NSC 97094, m-Chlorobenzoyl Hydroperoxide, m-Chloroperbenzoic Acid, m-Chloroperoxobenzoic Acid, m-Chloroperoxybenzoic Acid, m-Chloroperoxybenzoic Acid |
| InChI Formula | InChI=1S/C7H5ClO3/c8-6-3-1-2-5(4-6)7(9)11-10/h1-4,10H |
| SMILES | OOC(=O)c1cccc(Cl)c1 |
| IUPAC Name | 3-chlorobenzenecarboperoxoic acid |
| Molecular Weight (g/mol) | 172.57 |
| Formula Weight | 171.9927 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion