missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorophenol, TRC

Product Code. 30133012
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
Unit Size:
100g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133012 100 g 100g
17971435 25 g 25g
17922855 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133012 Supplier Toronto Research Chemicals Supplier No. TRCC375010100G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-43-0
Molecular Formula C6 H5 Cl O
Molecular Weight (g/mol) 128.56
Synonym Phenol, m-chloro- (7CI,8CI), 3-Chlorophenol, 3-Hydroxychlorobenzene, NSC 59700, m-Chlorophenic acid, m-Chlorophenol
IUPAC Name 3-chlorophenol
SMILES Oc1cccc(Cl)c1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C6H5ClO/c7-5-2-1-3-6(8)4-5/h1-4,8H
Formula Weight 128.0029
Chemical Name or Material 3-Chlorophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.