missing translation for 'onlineSavingsMsg'
Learn More

(3-Chloropropyl)benzene, 98%

Product Code. 11464407
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11464407

Brand: Thermo Scientific Alfa Aesar B20445.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-52-9 | C9H11Cl | 154.637 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-52-9
Molecular Formula C9H11Cl
Molecular Weight (g/mol) 154.637
MDL Number MFCD00001001
InChI Key XZBXAYCCBFTQHH-UHFFFAOYSA-N
Synonym 3-chloropropyl benzene, 1-chloro-3-phenylpropane, 3-phenylpropyl chloride, benzene, 3-chloropropyl, 3-phenyl-1-chloropropane, 3-chloro-1-phenylpropane, .gamma.-chloropropylbenzene, benzene, chloropropyl, .gamma.-phenylpropyl chloride, 3-penylpropylchloride
PubChem CID 7706
IUPAC Name 3-chloropropylbenzene
SMILES C1=CC=C(C=C1)CCCCl

Specifications

Density 1.05
Boiling Point 218°C to 220°C
Flash Point 87°C (189°F)
Linear Formula C6H5(CH2)3Cl
Refractive Index 1.522
Quantity 25 g
Formula Weight 154.64
Percent Purity 98%
Chemical Name or Material (3-Chloropropyl)benzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.