missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-5-hydroxypyridine, 97%

Product Code. 15405797
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15405797 250 mg 250mg
15495787 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15405797 Supplier Thermo Scientific Alfa Aesar Supplier No. H33006.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 152803-24-2 | C6H4N2O | 120.11 g/mol

3-Cyano-5-hydroxypyridine is active pharmaceutical ingredients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Cyano-5-hydroxypyridine is active pharmaceutical ingredients.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152803-24-2
Molecular Formula C6H4N2O
Molecular Weight (g/mol) 120.11
MDL Number MFCD11044307
InChI Key KAXIYYPIORYZLB-UHFFFAOYSA-N
PubChem CID 15342142
IUPAC Name 5-hydroxypyridine-3-carbonitrile
SMILES OC1=CN=CC(=C1)C#N

Specifications

Melting Point 213°C to 217°C
Quantity 250 mg
UN Number UN3439
Synonym 5-hydroxynicotinonitrile, 3-cyano-5-hydroxypyridine, 3-pyridinecarbonitrile, 5-hydroxy, 5-hydroxy-nicotinonitrile, 5-hydroxy-3-pyridinecarbonitrile, 5-cyano-3-hydroxypyridine, pubchem17727, kaxiyypioryzlb-uhfffaoysa, 5-cyano-5-hydroxypyridine, 3-pyridinecarbonitrile,5-hydroxy
Solubility Information Slightly soluble in water.
Formula Weight 120.11
Percent Purity 97%
Chemical Name or Material 3-Cyano-5-hydroxypyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.