missing translation for 'onlineSavingsMsg'
Learn More

3-Dechloro Sertraline Hydrochloride, TRC

Product Code. 17972495
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17972495 25 mg 25mg
17944475 50 mg 50mg
17963615 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17972495 Supplier Toronto Research Chemicals Supplier No. TRCD22665025MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79646-00-7
Molecular Formula C17H19Cl2N
Molecular Weight (g/mol) 308.25
Synonym (1S-cis)-4-(4-Chlorophenyl)-1,2,3,4-tetrahydro-N-methyl-1-naphthalenamine Hydrochloride;
IUPAC Name (1S,4S)-4-(4-chlorophenyl)-N-methyl-1,2,3,4-tetrahydronaphthalen-1-amine;hydrochloride
SMILES CN[C@H]1CC[C@H](C2=CC=CC=C12)C3=CC=C(C=C3)Cl.Cl

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C17H18ClN.ClH/c1-19-17-11-10-14(12-6-8-13(18)9-7-12)15-4-2-3-5-16(15)17;/h2-9,14,17,19H,10-11H2,1H3;1H/t14-,17-;/m0./s1
Formula Weight 307.0895
Chemical Name or Material 3-Dechloro Sertraline Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.