missing translation for 'onlineSavingsMsg'
Learn More

3-Descarboxy Olsalazine Methyl Ester, TRC

Product Code. 30367604
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367604 1 g 1g
30349062 250 mg 250mg
30350284 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30367604 Supplier Toronto Research Chemicals Supplier No. TRCD2215801G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Descarboxy Olsalazine Methyl Ester
Analyte or Component Names 3-Descarboxy Olsalazine Methyl Ester
Shipping Condition Room Temperature
Molecular Formula C14 H12 N2 O4
Synonym Methyl 2-Hydroxy-5-((4-hydroxyphenyl)diazenyl)benzoate
InChI Formula InChI=1S/C14H12N2O4/c1-20-14(19)12-8-10(4-7-13(12)18)16-15-9-2-5-11(17)6-3-9/h2-8,17-18H,1H3
Solution Type Neat
SMILES COC(=O)c1cc(ccc1O)N=Nc2ccc(O)cc2
IUPAC Name methyl 2-hydroxy-5-[(4-hydroxyphenyl)diazenyl]benzoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 272.256
Quantity 1 g
Formula Weight 272.08
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.