missing translation for 'onlineSavingsMsg'
Learn More

3-Descyano-3-formyl Febuxostat, TRC

Product Code. 30005328
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005328 250 mg 250mg
30012511 50 mg 50mg
30003666 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005328 Supplier Toronto Research Chemicals Supplier No. TRCD288145250MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144060-62-8
Molecular Formula C16 H17 N O4 S
Molecular Weight (g/mol) 319.38
Synonym Febuxostat Impurity 8
IUPAC Name 2-[3-formyl-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC(C)COc1ccc(cc1C=O)c2nc(C)c(s2)C(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C16H17NO4S/c1-9(2)8-21-13-5-4-11(6-12(13)7-18)15-17-10(3)14(22-15)16(19)20/h4-7,9H,8H2,1-3H3,(H,19,20)
Formula Weight 319.0878
Chemical Name or Material 2-[3-formyl-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.