missing translation for 'onlineSavingsMsg'
Learn More

3-(Di-n-butylamino)-1-propyne, 98%, Thermo Scientific™

Product Code. 11373026
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11373026 5 g 5g
11363026 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11373026 Supplier Thermo Scientific Alfa Aesar Supplier No. L10844.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6336-58-9
Molecular Formula C11H21N
Molecular Weight (g/mol) 167.296
MDL Number MFCD00041673
InChI Key XTEBRELWTGWYDE-UHFFFAOYSA-N
Synonym 3-dibutylamino-1-propyne, dibutylprop-2-ynylamine, dibutylpropargylamine, 3-dibutylaminopropyne, n,n-dibutylpropargylamine, di-n-butyl propargyl amine, acmc-209nf4, di-n-butylamino-1-propyne, n,n-dibutyl-2-propynylamine
PubChem CID 138732
IUPAC Name N-butyl-N-prop-2-ynylbutan-1-amine
SMILES CCCCN(CCCC)CC#C

Spécification

Boiling Point 87°C to 89°C (19 mmHg)
Odor Amine-like
Refractive Index 1.44
Quantity 5 g
Beilstein 1752173
Formula Weight 167.3
Percent Purity 98%
Chemical Name or Material 3-(Di-n-butylamino)-1-propyne
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.