missing translation for 'onlineSavingsMsg'
Learn More

3-(Diethylboryl)pyridine, TRC

Product Code. 30386113
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30386113 10 g 10g
30370651 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30386113 Supplier Toronto Research Chemicals Supplier No. TRCD44204010G
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl(3-pyridinyl)borane
CAS 89878-14-8
Molecular Formula C9 H14 B N
Quantity 10 g
Synonym 3-(Diethylboryl)pyridine, 3-Diethylboropyridine, 3-Pyridinyldiethylborane, 3-Pyridyldiethylborane, Diethyl(3-pyridinyl)borane, Diethyl-3-pyridylborane
InChI Formula InChI=1S/C9H14BN/c1-3-10(4-2)9-6-5-7-11-8-9/h5-8H,3-4H2,1-2H3
SMILES CCB(CC)c1cccnc1
IUPAC Name diethyl(pyridin-3-yl)borane
Molecular Weight (g/mol) 147.0252
Formula Weight 147.1219
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.