missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylaminobenzoyl chloride hydrochloride, tech. 90%

Product Code. 11305005
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11305005 1 g 1g
11315005 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11305005 Supplier Thermo Scientific Alfa Aesar Supplier No. L18433.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 117500-61-5 | C9H11Cl2NO | 220.093 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117500-61-5
Molecular Formula C9H11Cl2NO
Molecular Weight (g/mol) 220.093
MDL Number MFCD02094023
InChI Key NUDQPATXNFWFNK-UHFFFAOYSA-N
Synonym 3-dimethylaminobenzoyl chloride hydrochloride, 3-dimethylamino benzoyl chloride hydrochloride, acmc-20an50, 3-dimethylaminobenzoyl chloride hcl, 3-dimethylamino-benzoyl chloride hydrochloride salt, 3-dimethylamino benzoyl chloride-hydrogen chloride 1/1
PubChem CID 22732309
IUPAC Name 3-(dimethylamino)benzoyl chloride;hydrochloride
SMILES CN(C)C1=CC=CC(=C1)C(=O)Cl.Cl

Specifications

Melting Point 131°C to 133°C
Quantity 1 g
UN Number UN3261
Beilstein 2803337
Sensitivity Moisture sensitive
Formula Weight 220.1
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-Dimethylaminobenzoyl chloride hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.