missing translation for 'onlineSavingsMsg'
Learn More

3-(Dimethylphosphono)-N-methylolpropionamide (~70%), TRC

Product Code. 30133136
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133136 10 g 10g
30134317 5 g 5g
30012310 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133136 Supplier Toronto Research Chemicals Supplier No. TRCD48079810G
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20120-33-6
Molecular Formula C6H14NO5P
Molecular Weight (g/mol) 211.15
Synonym 3-(Dimethylphosphono)-N-(hydroxymethyl)propionamide;3-(Dimethylphosphono)-N-methylolpropionamide;3-(Dimethylphosphono)propionic acid methylolamide;3-(Dimethylphosphono)propionic methylolamide;3-(O,O'-Dimethylphosphono)-N-methylolpropionamide;Amgard TFR 1;CP;CP (flame retardant);CP New;Dimethyl N-methylolphosphonopropionamide;Dimethyl [2-[(hydroxymethyl)carbamoyl]ethyl]phosphonate;Dimethyl phosphonomethylolpropionamide;KX 912;N-Methylol-3-(dimethoxyphosphono)propionamide;N-Methylol-3-(dimethylphosphono)propionamide;Nicca Fi-None CG 700 Conc.;Pyrovatex 3805, Pyrovatex CF, Pyrovatex CP, Pyrovatex CP New, Spolapret OS
IUPAC Name 3-dimethoxyphosphoryl-N-(hydroxymethyl)propanamide
SMILES COP(=O)(CCC(=O)NCO)OC

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H14NO5P/c1-11-13(10,12-2)4-3-6(9)7-5-8/h8H,3-5H2,1-2H3,(H,7,9)
Formula Weight 211.061
Chemical Name or Material 3-(Dimethylphosphono)-N-methylolpropionamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.