missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-2-nitropyridine, 98%, Thermo Scientific™

Product Code. 10653532 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10653532 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10653532 Supplier Thermo Scientific Acros Supplier No. 152190010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 74037-50-6
CAS Min % 97.5
CAS Max % 100.0
Melting Point 28.0°C to 32.0°C
Boiling Point 175.0°C (50.0 mmHg)
Flash Point 38°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H8N2O3
MDL Number MFCD00006257
Quantity 1 g
Synonym pyridine, 3-ethoxy-2-nitro, 3-ethoxy-2-nitro-pyridine, 2-nitro-3-ethoxy pyridine, pubchem6624, acmc-209otd, 3-ethoxy-2-nitropyridin, 2-nitro-3-ethoxypyridine, pyridine,3-ethoxy-2-nitro, 4-21-00-00436 beilstein handbook reference
InChI Key WBEVIQNYYGPESH-UHFFFAOYSA-N
SMILES CCOC1=CC=CN=C1[N+]([O-])=O
IUPAC Name 3-ethoxy-2-nitropyridine
Molecular Weight (g/mol) 168.15
PubChem CID 52866
Formula Weight 168.15
Percent Purity 98%
Chemical Name or Material 3-Ethoxy-2-nitropyridine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Ethoxy-2-nitropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.