missing translation for 'onlineSavingsMsg'
Learn More

3-(Ethoxycarbonyl)phenyl isocyanate, 97%

Product Code. 11307466
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11307466 25 g 25g
11387456 1 g 1g
11397456 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11307466 Supplier Thermo Scientific Alfa Aesar Supplier No. L09495.14
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 67531-68-4 | C10H9NO3 | 191.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67531-68-4
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.19
MDL Number MFCD00013865
InChI Key UPXGGIRFDZENRD-UHFFFAOYSA-N
Synonym 3-ethoxycarbonyl phenyl isocyanate, 3-carbethoxyphenyl isocyanate, 3-ethoxycarbonylphenyl isocyanate, 3-ethoxycarbonyl phenylisocyanate, 3-isocyanatobenzoic acid ethyl ester, 3-carboethoxyphenyl isocyanate, #, ethyl 3-isocyanatobenzoate, 3-ethoxycarbonyl phenylisothiocyanate, 3-isocyanato-benzoic acid ethyl ester
PubChem CID 589806
IUPAC Name ethyl 3-isocyanatobenzoate
SMILES CCOC(=O)C1=CC(=CC=C1)N=C=O

Specifications

Density 1.154
Boiling Point 213°C
Flash Point >110°C (230°F)
Refractive Index 1.531
Quantity 25 g
UN Number UN2206
Beilstein 7636626
Sensitivity Moisture sensitive
Formula Weight 191.19
Percent Purity 97%
Chemical Name or Material 3-(Ethoxycarbonyl)phenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.