missing translation for 'onlineSavingsMsg'
Learn More

3-Ethylbenzophenone, TRC

Product Code. 30384689
Change view
Click to view available options
Quantity:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384689 25 mg
30368811 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384689 Supplier Toronto Research Chemicals Supplier No. TRCE89979025MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66067-43-4
Molecular Formula C15 H14 O
Molecular Weight (g/mol) 210.27
Synonym 3-Ethylbenzophenone, (3-Ethylphenyl)phenylmethanone, m-Ethylbenzophenone
IUPAC Name (3-ethylphenyl)-phenylmethanone
SMILES CCc1cccc(c1)C(=O)c2ccccc2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C15H14O/c1-2-12-7-6-10-14(11-12)15(16)13-8-4-3-5-9-13/h3-11H,2H2,1H3
Formula Weight 210.1045
Chemical Name or Material 3-Ethylbenzophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.