missing translation for 'onlineSavingsMsg'
Learn More

3-(Ethylsulfonyl)pyridine-2-sulfonamide, TRC

Product Code. 17982895
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17982895 500 mg 500mg
17932365 250 mg 250mg
17950335 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17982895 Supplier Toronto Research Chemicals Supplier No. TRCE926938500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117671-01-9
Molecular Formula C7 H10 N2 O4 S2
Molecular Weight (g/mol) 250.3
Synonym 3-(Ethylsulfonyl)-2-pyridinesulfonamide (ACI), 2-(Aminosulfonyl)-3-(ethylsulfonyl)pyridine, 2-Pyridinesulfonamide, 3-(ethylsulfonyl)- (9CI, ACI), IN-E9260
IUPAC Name 3-ethylsulfonylpyridine-2-sulfonamide
SMILES CCS(=O)(=O)c1cccnc1S(=O)(=O)N

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C7H10N2O4S2/c1-2-14(10,11)6-4-3-5-9-7(6)15(8,12)13/h3-5H,2H2,1H3,(H2,8,12,13)
Formula Weight 250.0082
Chemical Name or Material 3-(Ethylsulfonyl)pyridine-2-sulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.