Learn More
3-Fluoro-2-methylpyridine, 98%
Product Code. 15432019
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15432019 5 g 5g
15442019 250 mg 250mg
15422019 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15432019 Supplier Thermo Scientific Alfa Aesar Supplier No. H64705.06
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 15931-15-4 | C6H6FN | 111.119 g/mol
3-Fluoro-2-methylpyridine was used in the synthesis of fluorine-containing pyridinealdoximes of potential use for the reactivation of cholinesterase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Fluoro-2-methylpyridine was used in the synthesis of fluorine-containing pyridinealdoximes of potential use for the reactivation of cholinesterase.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15931-15-4
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.119
MDL Number MFCD08705710
InChI Key IRIHSZDBTACXCT-UHFFFAOYSA-N
Synonym 3-fluoro-2-picoline, pyridine, 3-fluoro-2-methyl, 2-methyl-3-fluoropyridine, 3-fluoro-2-methyl pyridine, 3-fluoro-2-methyl-pyridine, fluoropicoline, pubchem14381, acmc-209xci, ksc530a3b, pyridine,3-fluoro-2-methyl
PubChem CID 21613172
IUPAC Name 3-fluoro-2-methylpyridine
SMILES CC1=C(C=CC=N1)F
Specifications
Color Colorless to Yellow
Quantity 5 g
Beilstein 6497832
Solubility Information Slightly soluble in water.
Formula Weight 111.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-Fluoro-2-methylpyridine