missing translation for 'onlineSavingsMsg'
Learn More

3-Fluoro-4-methylbenzoyl chloride, 97%, Thermo Scientific™

Product Code. 10623993 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10623993 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10623993 Supplier Thermo Scientific Acros Supplier No. 375800010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 59189-97-8
CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C8H6ClFO
Linear Formula CH3C6H3(F)C(O)Cl
Refractive Index 1.5310 to 1.5330
MDL Number MFCD00671774
Quantity 1 g
Synonym 3-fluoro-4-methyl-benzoyl chloride, 3-fluoro-4-methylbenzoylchloride, acmc-20an3p, 3-fluoro-4-methyl-benzoylchloride, 3-fluoro-4-methyl benzoyl chloride, benzoyl chloride,3-fluoro-4-methyl, 3-fluoranyl-4-methyl-benzoyl chloride, benzoyl chloride, 3-fluoro-4-methyl, 3-fluoro-4-methylbenzoyl chloride, benzoyl chloride, 3-fluoro-4-methyl-9ci
Solubility Information Solubility in water: reacts
InChI Key DPWCZSXNEGNALT-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1F)C(Cl)=O
IUPAC Name 3-fluoro-4-methylbenzoyl chloride
Molecular Weight (g/mol) 172.58
PubChem CID 2734875
Formula Weight 172.59
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Fluoro-4-methylbenzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 3-Fluoro-4-methylbenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.