Learn More
3-Fluoro-4-methylphenol, 97%, Thermo Scientific™
Product Code. 15494517
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15494517 5 g 5g
15484517 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15494517 Supplier Thermo Scientific Alfa Aesar Supplier No. H31864.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 452-78-8
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00143164
InChI Key GJOOCAXPERKNMN-UHFFFAOYSA-N
Synonym 3-fluoro-p-cresol, 3-fluoro-4-methyl-phenol, 2-fluoro-4-hydroxytoluene, phenol, 3-fluoro-4-methyl, pubchem1499, 3-fluoro-4-methyl phenol, acmc-209k1q, 3-fluoranyl-4-methyl-phenol, ksc494a2d
PubChem CID 2774613
IUPAC Name 3-fluoro-4-methylphenol
SMILES CC1=C(C=C(C=C1)O)F
Specifications
Odor Phenol-like
Refractive Index 1.515
Quantity 5 g
UN Number UN3265
Formula Weight 126.13
Percent Purity 97%
Chemical Name or Material 3-Fluoro-4-methylphenol