missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Fluorophenmetrazine Hydrochloride (3-FPM HCl) 1.0 mg/ml in Methanol (as free base), LoGiCal
Specifications
Specifications
| Chemical Name or Material | 3-Fluorophenmetrazine Hydrochloride |
| CAS | 1803562-83-5 |
| Analyte or Component Names | 3-Fluorophenmetrazine Hydrochloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C11 H14 F N O . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Morpholine, 2-(3-fluorophenyl)-3-methyl-, hydrochloride (1:1), 3-Fluorophenmetrazine hydrochloride, 2-(3-Fluorophenyl)-3-methylmorpholine hydrochloride, 3-FPM HCl |
| InChI Formula | InChI=1S/C11H14FNO.ClH/c1-8-11(14-6-5-13-8)9-3-2-4-10(12)7-9;/h2-4,7-8,11,13H,5-6H2,1H3;1H |
| Solution Type | Single Solution |
| SMILES | Cl.CC1NCCOC1c2cccc(F)c2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?