missing translation for 'onlineSavingsMsg'
Learn More

3-Fluorophenyl isocyanate, 97%, Thermo Scientific™

Product Code. 10351144 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10351144 25 mL Glass bottle 25mL
10285740 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10351144 Supplier Thermo Scientific Acros Supplier No. 170330250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 404-71-7
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.11
MDL Number MFCD00002015
InChI Key RIKWVZGZRYDACA-UHFFFAOYSA-N
Synonym 3-fluorophenyl isocyanate, m-fluorophenyl isocyanate, 3-fluorophenylisocyanate, m-fluorophenylisocyanate, benzene, 1-fluoro-3-isocyanato, 3-isocyanatofluorobenzene, isocyanic acid 3-fluorophenyl ester, 3-fluorobenzenisocyanate, pubchem4995, acmc-1ajez
PubChem CID 123064
IUPAC Name 1-fluoro-3-isocyanatobenzene
SMILES FC1=CC=CC(=C1)N=C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.2010g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula FC6H4NCO
Refractive Index 1.5130 to 1.5150 (20°C, 589nm)
Quantity 25 mL
Formula Weight 137.11
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 3-Fluorophenyl isocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.