missing translation for 'onlineSavingsMsg'
Learn More

3-Furonitrile, 98%

Product Code. 11489830
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489830 5 g 5g
11398895 250 mg 250mg
11388895 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489830 Supplier Thermo Scientific Alfa Aesar Supplier No. L14019.06
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 30078-65-0 | C5H3NO | 93.085 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30078-65-0
Molecular Formula C5H3NO
Molecular Weight (g/mol) 93.085
MDL Number MFCD00067042
InChI Key ZQFYJHMUAWCEBH-UHFFFAOYSA-N
Synonym 3-furonitrile, 3-furancarbonitrile, 3-cyanofuran, 3-furancarbonitrile 9ci, 3-furonitrile 8ci, acmc-1cj8r
PubChem CID 100698
IUPAC Name furan-3-carbonitrile
SMILES C1=COC=C1C#N

Specifications

Melting Point 28°C to 30°C
Boiling Point 150°C to 151°C
Flash Point 53°C (127°F)
Refractive Index 1.479
Quantity 5 g
UN Number UN2926
Beilstein 507009
Formula Weight 93.09
Percent Purity 98%
Chemical Name or Material 3-Furonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.