Learn More
3-Heptyn-1-ol, 98%
Product Code. 11449916
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449916 5 g 5g
11459916 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11449916 Supplier Thermo Scientific Alfa Aesar Supplier No. B20884.06
Request Bulk or Custom Format
CAS: 14916-79-1 | C7H12O | 112.172 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14916-79-1
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.172
MDL Number MFCD00039551
InChI Key PSWHODJVUOXHKA-UHFFFAOYSA-N
Synonym 3-heptyn-1-ol, acmc-209d1z, ho ch2 2c=c ch2 2ch3
PubChem CID 84693
IUPAC Name hept-3-yn-1-ol
SMILES CCCC#CCCO
Specifications
Density 0.882
Boiling Point 80°C to 82°C (15 mmHg)
Flash Point 74°C (165°F)
Refractive Index 1.4555
Quantity 5 g
Beilstein 1737178
Formula Weight 112.17
Percent Purity 98%
Chemical Name or Material 3-Heptyn-1-ol