missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hexanone-2,2,4,4-d4, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 3-Hexanone-2,2,4,4-D4 |
| CAS | 24588-54-3 |
| Molecular Formula | C6 D4 H8 O |
| Quantity | 0.5 g |
| Synonym | 3-Hexanone-2,2,4,4-d4 (8 CI, 9 CI), 3-Hexanone-2,2,4,4-D4 |
| InChI Formula | InChI=1 S/C6H12O/c1-3-5-6(7)4-2/h3-5H2,1-2H3/i4D2,5D2 |
| SMILES | [2 H]C([2 H])(C)C(=O)C([2 H])([2 H])CC |
| IUPAC Name | 2,2,4,4-tetradeuteriohexan-3-one |
| Molecular Weight (g/mol) | 104.1835 |
| Formula Weight | 104.1139 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction