missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-1-methylpyrrolidine, 97%

Product Code. 11357599
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357599 5 g 5g
11347599 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357599 Supplier Thermo Scientific Alfa Aesar Supplier No. H28122.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13220-33-2 | C5H11NO | 101.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13220-33-2
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00003176
InChI Key FLVFPAIGVBQGET-UHFFFAOYSA-N
Synonym 1-methyl-3-pyrrolidinol, 3-hydroxy-1-methylpyrrolidine, n-methyl-3-pyrrolidinol, 3-pyrrolidinol, 1-methyl, 3-hydroxy-n-methylpyrrolidine, n-methyl-3-hydroxypyrrolidine, 1-methyl-3-hydroxypyrrolidine, pubchem14117, l-methyl-3-pyrrolidinol, 1-methyl-pyrrolidin-3ol
PubChem CID 93074
IUPAC Name 1-methylpyrrolidin-3-ol
SMILES CN1CCC(C1)O

Specifications

Density 0.921
Boiling Point 50°C to 52°C (1 mmHg)
Flash Point 71°C (160°F)
Refractive Index 1.464
Quantity 5 g
Formula Weight 101.15
Percent Purity 97%
Chemical Name or Material 3-Hydroxy-1-methylpyrrolidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.