missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy-2,2-dimethylpropionic acid neopentyl glycol ester, Thermo Scientific™

Product Code. 11425750
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425750 500 g 500g
11415750 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425750 Supplier Thermo Scientific Alfa Aesar Supplier No. H25963.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Hydroxy-2,2-dimethylpropionic acid neopentyl glycol ester is a reactant in the Tischenko Reaction.

Solubility
Soluble in methanol(almost transparency).

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1115-20-4
Molecular Formula C10H20O4
Molecular Weight (g/mol) 204.266
MDL Number MFCD00059597
InChI Key SZCWBURCISJFEZ-UHFFFAOYSA-N
Synonym hydroxypivalic acid neopentyl glycol ester, esterdiol 204, 3-hydroxy-2,2-dimethylpropyl 3-hydroxy-2,2-dimethylpropionate, hydroxyneopentyl hydroxypivalate, unii-cg0a37gahj, propanoic acid, 3-hydroxy-2,2-dimethyl-, 3-hydroxy-2,2-dimethylpropyl ester, neopentyl glycol monohydroxypivalate, 3-hydroxypivaloyloxy-2,2-dimethylpropanol, 3-hydroxy-2,2-dimethylpropyl hydroxypivalate, neopental glycol monohydroxypivalate
PubChem CID 14218
IUPAC Name (3-hydroxy-2,2-dimethylpropyl) 3-hydroxy-2,2-dimethylpropanoate
SMILES CC(C)(CO)COC(=O)C(C)(C)CO

Spezifikation

Melting Point 51°C to 54°C
Density 1.01
Boiling Point 292°C
Flash Point 161°C (322°F)
Quantity 500 g
Sensitivity Hygroscopic
Solubility Information Soluble in methanol(almost transparency).
Formula Weight 204.27
Chemical Name or Material 3-Hydroxy-2,2-dimethylpropionic acid neopentyl glycol ester
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.