missing translation for 'onlineSavingsMsg'
Learn More

3-Hydroxy Benzopyrene, TRC

Product Code. 17933605
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17933605 5 mg 5mg
17994115 1 mg 1mg
17904765 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17933605 Supplier Toronto Research Chemicals Supplier No. TRCH8294005MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13345-21-6
Molecular Formula C20 H12 O
Molecular Weight (g/mol) 268.31
Synonym Benzo[a]pyren-3-ol, 3-Hydroxy-3,4-benzo[a]pyrene, 3-Hydroxybenzo[a]pyrene;
IUPAC Name benzo[a]pyren-3-ol
SMILES Oc1ccc2ccc3c4ccccc4cc5ccc1c2c35

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C20H12O/c21-18-10-7-12-5-8-16-15-4-2-1-3-13(15)11-14-6-9-17(18)19(12)20(14)16/h1-11,21H
Formula Weight 268.0888
Percent Purity >95%
Chemical Name or Material 3-Hydroxy Benzopyrene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.