Learn More
3-Hydroxymethylantipyrine, TRC
Product Code. 30136261
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136261 100 mg 100mg
30132541 250 mg 250mg
30005198 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30136261 Supplier Toronto Research Chemicals Supplier No. TRCH805090100MG
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 18125-49-0
Molecular Formula C11H12N2O2
Molecular Weight (g/mol) 204.23
Synonym 3-(Hydroxymethyl)-2-methyl-1-phenyl-3-pyrazolin-5-one, HMA, 1,2-Dihydro-5-(hydroxymethyl)-1-methyl-2-phenyl-3H-pyrazol-3-one, 3-(Hydroxymethyl)-2-methyl-1-phenyl-3-pyrazolin-5-one, Hydroxymethylantipyrine
IUPAC Name 5-(hydroxymethyl)-1-methyl-2-phenylpyrazol-3-one
SMILES CN1N(C(=O)C=C1CO)c2ccccc2
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C11H12N2O2/c1-12-10(8-14)7-11(15)13(12)9-5-3-2-4-6-9/h2-7,14H,8H2,1H3
Formula Weight 204.0899
Chemical Name or Material 3-Hydroxymethylantipyrine