missing translation for 'onlineSavingsMsg'
Learn More

3'-Hydroxypuerarin, TRC

Product Code. 30180117
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30180117 10 mg 10mg
30180008 1 mg 1mg
30179817 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180117 Supplier Toronto Research Chemicals Supplier No. TRCH95404510MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117060-54-5
Molecular Formula C21 H20 O10
Molecular Weight (g/mol) 432.38
Synonym 4H-1-Benzopyran-4-one, 3-(3,4-dihydroxyphenyl)-8-β-D-glucopyranosyl-7-hydroxy-, 3-(3,4-Dihydroxyphenyl)-8-β-D-glucopyranosyl-7-hydroxy-4H-1-benzopyran-4-one, 3'-Hydroxypuerarin, 8-C-Glucosyl-7,3',4'-trihydroxyisoflavone, NPI 031H, PG 1
IUPAC Name 3-(3,4-dihydroxyphenyl)-7-hydroxy-8-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]chromen-4-one
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)ccc3C(=O)C(=COc23)c4ccc(O)c(O)c4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H20O10/c22-6-14-17(27)18(28)19(29)21(31-14)15-12(24)4-2-9-16(26)10(7-30-20(9)15)8-1-3-11(23)13(25)5-8/h1-5,7,14,17-19,21-25,27-29H,6H2/t14-,17-,18+,19-,21+/m1/s1
Formula Weight 432.1056
Chemical Name or Material 3'-Hydroxypuerarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.