missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Iodo-4-(2,2,2-trimethylacetamido)pyridine

Product Code. 11404400
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11404400

Brand: Alfa Aesar™ H50026.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 113975-33-0
Molecular Formula C10H13IN2O
Molecular Weight (g/mol) 304.131
MDL Number MFCD04971323
InChI Key GPMKCDBJLNTANL-UHFFFAOYSA-N
Synonym n-3-iodopyridin-4-yl pivalamide, n-3-iodo-pyridin-4-yl-2,2-dimethyl-propionamide, n-3-iodopyridin-4-yl-2,2-dimethylpropanamide, n-3-iodo-4-pyridinyl-2,2-dimethylpropanamide, 3-iodo-4-2,2,2-trimethylacetamido pyridine, n-3-iodo-4-pyriidinyl-2,2-dimethylpropanamide, propanamide, n-3-iodo-4-pyridinyl-2,2-dimethyl, acmc-20akbz, n-3-iodo-4-pyridyl pivalamide, n-3-iodopyridin-4-yl-2,2-dimethylpropionamide
PubChem CID 819120
IUPAC Name N-(3-iodopyridin-4-yl)-2,2-dimethylpropanamide
SMILES CC(C)(C)C(=O)NC1=C(C=NC=C1)I

Specifications

Quantity 5 g
Sensitivity Light sensitive
Formula Weight 304.13
Chemical Name or Material 3-Iodo-4-(2,2,2-trimethylacetamido)pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.