missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-4-methylbenzoic acid, 98%

Product Code. p-7021739
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11412277

Brand: Thermo Scientific Alfa Aesar A14197.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 82998-57-0 | C8H7IO2 | 262.046 g/mol

3-Iodo-4-methylbenzoic acid has been used in the preparation of unlabeled N-succinimidyl 4-guanidinomethyl-3-iodobenzoate (SGIMB) and boc-protected derivative of SGIMB (Boc-SGMIB).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Iodo-4-methylbenzoic acid has been used in the preparation of unlabeled N-succinimidyl 4-guanidinomethyl-3-iodobenzoate (SGIMB) and boc-protected derivative of SGIMB (Boc-SGMIB).

Solubility
Soluble in methanol.

Notes
Recommended storage temp 2-8°C. Store in cool, well ventilated area. Keep container tightly closed. Light Sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82998-57-0
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
MDL Number MFCD00010392
InChI Key LDDHMKANNXWUAK-UHFFFAOYSA-N
Synonym 3-iodo-4-methyl-benzoic acid, benzoic acid, 3-iodo-4-methyl, zlchem 953, 3-iodo-p-toluicacid, pubchem3947, 4-carboxy-2-iodotoluene, akos bbb/227, ksc449e9t, 3-iodo-4-methyl benzoic acid, paragos 440021
PubChem CID 621640
IUPAC Name 3-iodo-4-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)I

Specifications

Melting Point 206°C to 210°C
Quantity 5 g
Beilstein 2436216
Sensitivity Light sensitive
Solubility Information Soluble in methanol.
Formula Weight 262.05
Percent Purity 98%
Chemical Name or Material 3-Iodo-4-methylbenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.