missing translation for 'onlineSavingsMsg'
Learn More

3-Iodobenzeneboronic acid pinacol ester, 97%

Product Code. 11377589
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377589 1 g 1g
11387589 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377589 Supplier Thermo Scientific Alfa Aesar Supplier No. H28115.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 408492-28-4 | C12H16BIO2 | 329.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 408492-28-4
Molecular Formula C12H16BIO2
Molecular Weight (g/mol) 329.97
MDL Number MFCD09953498
InChI Key HSHFNMSHDHAHDN-UHFFFAOYSA-N
Synonym 2-3-iodophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-iodophenylboronic acid pinacol ester, amtb088, 3-iodophenylboronic acid pinacolester, 1,3,2-dioxaborolane,2-3-iodophenyl-4,4,5,5-tetramethyl, 2-3-iodanylphenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-iodo-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzene
PubChem CID 11313483
IUPAC Name 2-(3-iodophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC(I)=CC=C1

Specifications

Melting Point 70°C to 72°C
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 329.97
Percent Purity 97%
Chemical Name or Material 3-Iodobenzeneboronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.