Learn More
3-Mercapto-1,2,4-triazole, 98%
Product Code. 11404474
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11404474 5 g 5g
11414474 25 g 25g
11424474 100 g 100g
3 options
Product Code. 11404474
Brand: Thermo Scientific Alfa Aesar A13120.06
Request Bulk or Custom Format
CAS: 3179-31-5 | C2H3N3S | 101.13 g/mol
3-Mercapto-1,2,4-triazole was used in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Mercapto-1,2,4-triazole was used in a study to design a surface enhanced Raman scattering based probe for fast and accurate detection of DNA markers.
Solubility
Soluble in hot water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3179-31-5
Molecular Formula C2H3N3S
Molecular Weight (g/mol) 101.13
MDL Number MFCD00005229
InChI Key AFBBKYQYNPNMAT-UHFFFAOYSA-N
Synonym 1h-1,2,4-triazole-3-thiol, 3-mercapto-1,2,4-triazole, 1,2,4-triazole-3-thiol, mercaptotriazole, 4h-1,2,4-triazole-3-thiol, s-triazole-3-thiol, 3-mercapto-1h-1,2,4-triazole, 1,2,4-triazole-5-thiol, 1,3,4-triazine-2-thiol, 1,3,4-triazole-2-thiol
PubChem CID 2723802
SMILES S=C1NNC=N1
Specifications
Melting Point 217°C to 224°C
Quantity 5 g
Beilstein 107731
Solubility Information Soluble in hot water.
IUPAC Name 1,2-dihydro-1,2,4-triazole-3-thione
Formula Weight 101.13
Percent Purity 98%
Chemical Name or Material 3-Mercapto-1,2,4-triazole