missing translation for 'onlineSavingsMsg'
Learn More

3-Mercaptopropionic Acid, TRC

Product Code. 30465992
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465992 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30465992 Supplier Toronto Research Chemicals Supplier No. TRCM2570701G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Mercaptopropionic Acid
CAS 107-96-0
Molecular Formula C3 H6 O2 S
Quantity 1 g
Synonym Propanoic acid, 3-mercapto- (9CI, ACI), 3-Mercaptopropanoic acid (ACI), Propionic acid, 3-mercapto- (8CI), Propionic acid, β-mercapto- (4CI), 2-Mercaptoethanecarboxylic acid, 3-Mercaptopropionic acid, 3-Sulfanylpropanoic acid, 3-Thiopropanoic acid, 3-Thiopropionic acid, BMPA, BMPA (thiol), Mercaptopropionic acid, MeSH ID: D015097, MPA, NSC 437, NSC 45157, R 013140, Thiohydracrylic acid, β-Mercaptopropanoic acid, β-Mercaptopropionic acid, β-Thiopropionic acid
InChI Formula InChI=1S/C3H6O2S/c4-3(5)1-2-6/h6H,1-2H2,(H,4,5)
SMILES OC(=O)CCS
IUPAC Name 3-sulfanylpropanoic acid
Molecular Weight (g/mol) 106.14
Formula Weight 106.0089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.