Learn More
3-Methyl-2-(1-piperazinyl)quinoline, 97%, Thermo Scientific™
Product Code. 11414850
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11414850 250 mg 250mg
11424850 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11414850 Supplier Thermo Scientific Alfa Aesar Supplier No. H50834.MD
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 348133-74-4
Molecular Formula C14H17N3
Molecular Weight (g/mol) 227.311
MDL Number MFCD08061017
InChI Key YUKANULGPVFVJO-UHFFFAOYSA-N
Synonym 3-methyl-2-piperazin-1-yl quinoline, 3-methyl-2-1-piperazinyl quinoline, 3-methyl-2-piperazin-1-yl-quinoline, acmc-20an0s, 3-methyl-2-quinolyl piperazine, 3-methyl-2-piperazine-1-yl quinoline
PubChem CID 22636341
IUPAC Name 3-methyl-2-piperazin-1-ylquinoline
SMILES CC1=CC2=CC=CC=C2N=C1N3CCNCC3
Specifications
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 227.31
Percent Purity 97%
Chemical Name or Material 3-Methyl-2-(1-piperazinyl)quinoline