missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-butanone, TRC

Product Code. 30465172
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30465172 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30465172 Supplier Toronto Research Chemicals Supplier No. TRCM294155100ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Methyl-2-butanone
CAS 563-80-4
Molecular Formula C5 H10 O
Quantity 100 mL
Synonym 2-Butanone, 3-methyl-, 3-Methyl-2-butanone, 2-Acetylpropane, 2-Methylbutan-3-one, Isopropyl methyl ketone, MIPK, Methyl isopropyl ketone, NSC 9379
InChI Formula InChI=1S/C5H10O/c1-4(2)5(3)6/h4H,1-3H3
SMILES CC(C)C(=O)C
IUPAC Name 3-methylbutan-2-one
Molecular Weight (g/mol) 86.13
Formula Weight 86.0732
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.