missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-5-phenyl-1H-pyrazole, Thermo Scientific Chemicals

Product Code. 11490170
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11490170 1 g 1g
11400180 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490170 Supplier Thermo Scientific Alfa Aesar Supplier No. H50650.03
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3347-62-4 | Molecular Formula: C10H10N2 | Molecular Weight: 158.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3347-62-4
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.20
MDL Number MFCD00022385,MFCD00524752
InChI Key QHRSESMSOJZMCO-UHFFFAOYSA-N
PubChem CID 18774
IUPAC Name 5-methyl-3-phenyl-1H-pyrazole
SMILES CC1=CC(=NN1)C1=CC=CC=C1

Specifications

Melting Point 124°C to 127°C
Quantity 1 g
Synonym 3-methyl-5-phenyl-1h-pyrazole, femerazol, 3-methyl-5-phenylpyrazole, phemerazole, 1h-pyrazole, 3-methyl-5-phenyl, 5-methyl-3-phenylpyrazole, 1h-pyrazole,3-methyl-5-phenyl, pyrazole, 3-methyl-5-phenyl, unii-0xfx19d77a, 1h-pyrazole, 5-methyl-3-phenyl
Formula Weight 158.2
Physical Form Powder
Chemical Name or Material 3-Methyl-5-phenyl-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.