Learn More
3-Methyl-7-propylxanthine, Mikromol™
Product Code. 30049836
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049836 25 mg 25mg
30049065 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30049836 Supplier Mikromol Supplier No. MM0875.010025
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3-Methyl-7-propylxanthine
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Methyl-7-propylxanthine
CAS 55242-64-3
Analyte or Component Names 3-Methyl-7-propylxanthine
Shipping Condition Room Temperature
Molecular Formula C9 H12 N4 O2
Certifications/Compliance ISO 9001
Synonym 3-Methyl-7-propylxanthine
InChI Formula InChI=1S/C9H12N4O2/c1-3-4-13-5-10-7-6(13)8(14)11-9(15)12(7)2/h5H,3-4H2,1-2H3,(H,11,14,15)
SMILES CCCn1cnc2N(C)C(=O)NC(=O)c12
IUPAC Name 3-methyl-7-propylpurine-2,6-dione
Molecular Weight (g/mol) 208.22
Quantity 25 mg
Formula Weight 208.096 g/mol