Learn More
3-Methyl-7-propylxanthine, TRC
Product Code. 30137127
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137127 1 g 1g
30008528 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30137127 Supplier Toronto Research Chemicals Supplier No. TRCM3260131G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 55242-64-3
Molecular Formula C9 H12 N4 O2
Molecular Weight (g/mol) 208.22
Synonym 3-Methyl-7-propylxanthine, 3,7-Dihydro-3-methyl-7-propyl-1H-purine-2,6-dione, 3-Methyl-7-n-propylxanthine;
IUPAC Name 3-methyl-7-propylpurine-2,6-dione
SMILES CCCn1cnc2N(C)C(=O)NC(=O)c12
Specifications
Quantity 1 g
InChI Formula InChI=1S/C9H12N4O2/c1-3-4-13-5-10-7-6(13)8(14)11-9(15)12(7)2/h5H,3-4H2,1-2H3,(H,11,14,15)
Formula Weight 208.096
Chemical Name or Material 3-Methyl-7-propylxanthine