Learn More
Alfa Aesar™ 3-Methyladipic acid, 99%, Thermo Scientific™
Product Code. 11439337
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439337 100 g 100g
11429337 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11439337 Supplier Alfa Aesar™ Supplier No. B21387.22
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3058-01-3
MDL Number MFCD00002804
ChEBI CHEBI:68503
Specifications
Melting Point 97°C to 102°C
Boiling Point 229°C to 231°C (30 mmHg)
Molecular Formula C7H12O4
Quantity 100 g
Synonym 3-methyladipic acid, hexanedioic acid, 3-methyl, +-3-methyladipic acid, beta-methyl-adipic acid, 3-methyl-hexanedioic acid, 3-methyl adipic acid, 3-methyl-adipic acid, acmc-20ah5v, acmc-1cr8a, hexanedioic acid,3-methyl
InChI Key SYEOWUNSTUDKGM-UHFFFAOYNA-N
SMILES CC(CCC(=O)O)CC(=O)O
IUPAC Name 3-methylhexanedioic acid
Molecular Weight (g/mol) 160.17
PubChem CID 12292
Formula Weight 160.17
Percent Purity 99%
Chemical Name or Material 3-Methyladipic acid