missing translation for 'onlineSavingsMsg'
Learn More

3-Methylbenzofuran-2-carboxylic acid, 98%, ACROS Organics™

Product Code. 10580902 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10580902 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10580902 Supplier Thermo Scientific Acros Supplier No. 356360010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 24673-56-1
CAS Min % 97.5
CAS Max % 100.0
Melting Point 192.0°C to 197.0°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C10H8O3
MDL Number MFCD00060511
Quantity 1 g
Synonym 3-methylbenzofuran-2-carboxylic acid, 2-benzofurancarboxylic acid, 3-methyl, 3-methyl-2-benzofurancarboxylic acid, 3-methyl-benzofuran-2-carboxylic acid, 3-methylbenzo b furan-2-carboxylic acid, cambridge id 5100668, acmc-1cp97, 3-methylbenzofuran-2-carboxylicacid, 3-methylbenzofuran 2 carboxylic acid, 2-benzofurancarboxylicacid, 3-methyl
InChI Key YMZTUCZCQMQFMK-UHFFFAOYSA-N
SMILES CC1=C(OC2=CC=CC=C12)C(=O)O
IUPAC Name 3-methyl-1-benzofuran-2-carboxylic acid
Molecular Weight (g/mol) 176.171
PubChem CID 600591
Formula Weight 176.17
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3-Methylbenzofuran-2-carboxylic acid, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.