missing translation for 'onlineSavingsMsg'
Learn More

3-Methylindole, TRC

Product Code. 30470915
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 30470915

Brand: Toronto Research Chemicals TRCM313470100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Methylindole
CAS 83-34-1
Molecular Formula C9 H9 N
Quantity 100 g
Synonym 1H-Indole','3-methyl-, Indole','3-methyl- (8CI), Indole','3-methyl-(skatole) (7CI), 3-Methyl-1H-indole, 3-Methylindole, NSC 122024, Scatole, Skatol, Skatole, β-Methylindole
InChI Formula InChI=1S/C9H9N/c1-7-6-10-9-5-3-2-4-8(7)9/h2-6,10H,1H3
SMILES Cc1c[nH]c2ccccc12
IUPAC Name 3-methyl-1H-indole
Molecular Weight (g/mol) 131.17
Formula Weight 131.0735
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.