missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methylindole, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 3-Methylindole |
| CAS | 83-34-1 |
| Molecular Formula | C9 H9 N |
| Quantity | 10 g |
| Synonym | 1H-Indole','3-methyl-, Indole','3-methyl- (8CI), Indole','3-methyl-(skatole) (7CI), 3-Methyl-1H-indole, 3-Methylindole, NSC 122024, Scatole, Skatol, Skatole, β-Methylindole |
| InChI Formula | InChI=1S/C9H9N/c1-7-6-10-9-5-3-2-4-8(7)9/h2-6,10H,1H3 |
| SMILES | Cc1c[nH]c2ccccc12 |
| IUPAC Name | 3-methyl-1H-indole |
| Molecular Weight (g/mol) | 131.17 |
| Formula Weight | 131.0735 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction