Learn More
3-Methylquinoline, 98+%
Product Code. 11424454
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11424454 25 g 25g
11414454 5 g 5g
11434454 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11424454 Supplier Thermo Scientific Alfa Aesar Supplier No. A13109.14
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 612-58-8 | C10H9N | 143.19 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 612-58-8
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00014661
InChI Key DTBDAFLSBDGPEA-UHFFFAOYSA-N
Synonym quinoline, 3-methyl, 3-methyl-1-benzazine, 3-methyl-quinoline, ccris 2896, unii-1yc9rbw2g0, beta-methylquinoline, 1yc9rbw2g0, 3-methyl quinoline, pubchem2344, acmc-209mql
PubChem CID 11926
ChEBI CHEBI:20140
IUPAC Name 3-methylquinoline
SMILES CC1=CN=C2C=CC=CC2=C1
Specifications
Melting Point 15°C
Density 1.069
Boiling Point 252°C to 253°C
Flash Point >110°C (230°F)
Refractive Index 1.615
Quantity 25 g
Beilstein 110325
Formula Weight 143.19
Percent Purity ≥98%
Chemical Name or Material 3-Methylquinoline