missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)aniline, 97%, ACROS Organics™

Product Code. 10468890 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10468890 10 g Glass bottle 10g
10574101 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10468890 Supplier Thermo Scientific Acros Supplier No. 163190100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1783-81-9
Molecular Formula C7H9NS
Molecular Weight (g/mol) 139.216
MDL Number MFCD00007793
InChI Key KCHLDNLIJVSRPK-UHFFFAOYSA-N
Synonym 3-methylthio aniline, 3-aminothioanisole, m-aminothioanisole, m-methylthio aniline, benzenamine, 3-methylthio, 3-methylmercapto aniline, 3-methylsulfanyl aniline, 3-methylmercaptoaniline, 3-methylthiobenzenamine, m-aminophenyl methyl sulfide
PubChem CID 15693
IUPAC Name 3-methylsulfanylaniline
SMILES CSC1=CC=CC(=C1)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Brown
Density 1.1300g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3SC6H4NH2
Refractive Index 1.6410 to 1.6450 (20°C, 589nm)
Quantity 10 g
Beilstein 13, IV, 1007
Formula Weight 139.21
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 3-(Methylthio)aniline, 97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.