missing translation for 'onlineSavingsMsg'
Learn More

3-(Methylthio)aniline, 97%

Product Code. 11460777
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460777 5 g 5g
11369549 25 g 25g
11379549 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460777 Supplier Thermo Scientific Alfa Aesar Supplier No. B23184.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1783-81-9 | C7H9NS | 139.216 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1783-81-9
Molecular Formula C7H9NS
Molecular Weight (g/mol) 139.216
MDL Number MFCD00007793
InChI Key KCHLDNLIJVSRPK-UHFFFAOYSA-N
Synonym 3-methylthio aniline, 3-aminothioanisole, m-aminothioanisole, m-methylthio aniline, benzenamine, 3-methylthio, 3-methylmercapto aniline, 3-methylsulfanyl aniline, 3-methylmercaptoaniline, 3-methylthiobenzenamine, m-aminophenyl methyl sulfide
PubChem CID 15693
IUPAC Name 3-methylsulfanylaniline
SMILES CSC1=CC=CC(=C1)N

Specifications

Density 1.13
Boiling Point 163°C to 165°C (16 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Refractive Index 1.637
Quantity 5 g
UN Number UN2810
Beilstein 2078599
Formula Weight 139.23
Percent Purity 97%
Chemical Name or Material 3-(Methylthio)aniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.