missing translation for 'onlineSavingsMsg'
Learn More

3-Methylthiophene, TRC

Product Code. 30137838
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137838 10 g 10g
30004003 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137838 Supplier Toronto Research Chemicals Supplier No. TRCM33026510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-44-4
Molecular Formula C5 H6 S
Molecular Weight (g/mol) 98.17
Synonym 3-Methylthiophene, 3-Methylthiophene (ACI), 3-Thienylmethane, 3-Thiotolene, NSC 65439, β-Methylthiophene, 3-Methylthiophene, 3-Methylthiophene (ACI), 3-Thienylmethane, 3-Thiotolene, NSC 65439, β-Methylthiophene
IUPAC Name 3-methylthiophene
SMILES Cc1ccsc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C5H6S/c1-5-2-3-6-4-5/h2-4H,1H3
Formula Weight 98.019
Chemical Name or Material 3-Methylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.