missing translation for 'onlineSavingsMsg'
Learn More

3-(N,N-Dimethylpalmitylammonio)propanesulfonate, TRC

Product Code. 30384681
Change view
Click to view available options
Quantity:
25 g
50 g
Unit Size:
25g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30384681 50 g 50g
30377512 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30384681 Supplier Toronto Research Chemicals Supplier No. TRCD46182550G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(N,N-Dimethylpalmitylammonio)propanesulfonate
CAS 2281-11-0
Molecular Formula C21H45NO3S
Quantity 50 g
Synonym N,N-Dimethyl-N-(3-sulfopropyl)-1-hexadecanaminium Hydroxide Inner Salt, Hexadecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt, Hexadecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt, 3-(Dimethylhexadecylammonio)propanesulfonate, 3-(Hexadecyldimethylammonio)propane-1-sulfonate, 3-(Hexadecyldimethylammonio)propanesulfonate, 3-(Hexadecyldimethylammonium)-1-sulfopropane Betaine, 3-(N,N-Dimethyl-N-hexadecylammonio)propane-1-sulfonate, 3-(N,N-Dimethylpalmitylammonio)propanesulfonate, 3-(N,NDimethylpalmitylammonio)
InChI Formula InChI=1S/C21H45NO3S/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22(2,3)20-18-21-26(23,24)25/h4-21H2,1-3H3
SMILES [O-]S(CCC[N+](C)(C)CCCCCCCCCCCCCCCC)(=O)=O
IUPAC Name 3-(hexadecyldimethylammonio)propane-1-sulfonate
Molecular Weight (g/mol) 391.65
Formula Weight 391.31
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.