Learn More
Eppendorf™ Research 3 Neo Single Channel Pipettes Piston
Product Code. 30689725 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
2.5 μL
10 μL
20 μL
100 μL
200 μL
300 μL
1000 μL
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
30689725 100 μL Each
30689603 2.5 μL Each
30689624 10 μL Each
30689716 20 μL Each
30689607 200 μL Each
30689656 300 μL Each
30689714 1000 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 30689725 Supplier Eppendorf™ Supplier No. 3102633004
Spotlight Offers
Use promo code 28037
View all promotions
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Restore the precision of your Eppendorf mechanical pipette with long-lasting performance
  • Designed for smooth operation and long-lasting performance
  • This spare part guarantees precise control and consistent results
  • Restore the precision of your Eppendorf mechanical pipette with an original replacement for a worn or damaged piston
TRUSTED_SUSTAINABILITY
Specifications
For Use With Pipettes
Material Thermoplastic
Type Piston
Volume (Metric) 100 μL
For Use With (Equipment) Pipette Tips
Quantity 1 Pk.